missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexabromotellurate(IV), 99.99% (metals basis), Thermo Scientific™

Product Code. 11315178
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315178 5 g 5g
11395168 25 g 25g
11305178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11315178 Supplier Thermo Scientific Alfa Aesar Supplier No. 044081.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16986-18-8
Molecular Formula Br6K2Te
Molecular Weight (g/mol) 685.221
MDL Number MFCD03427026
InChI Key HMFRPYOIJYYQFO-UHFFFAOYSA-L
Synonym potassium hexabromotellurate iv
PubChem CID 121233754
SMILES [K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 5 g
UN Number UN3284
Formula Weight 685.22
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Potassium hexabromotellurate (IV)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.