missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexabromoiridate(IV), 99.9% (metals basis), Ir 25.2% min

Product Code. 11309417
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309417 1 g 1g
11319417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309417 Supplier Thermo Scientific Alfa Aesar Supplier No. 041012.03
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19121-78-9 | Br6IrK2 | 749.838 g/mol

It is a precious metal compound and a homogeneous catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19121-78-9
Molecular Formula Br6IrK2
Molecular Weight (g/mol) 749.838
MDL Number MFCD00054221
InChI Key RSDYABYRTHADQA-UHFFFAOYSA-H
Synonym potassium hexabromoiridate iv, dipotassium;hexabromoiridium 2-, dipotassium hexabromoiridiumdiuide, potassium hexabromoiridate iv , ir
PubChem CID 22555925
IUPAC Name dipotassium;hexabromoiridium(2-)
SMILES [K+].[K+].Br[Ir-2](Br)(Br)(Br)(Br)Br

Specifications

Color Black
Physical Form Crystalline
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 749.86
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ir 25.2% min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Potassium hexabromoiridate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.