Learn More
Potassium heptafluorotantalate(V), 99.99% (metals basis)
Description
It is an important raw material used as a primary and secondary pharmaceutical intermediate and also used in the production and preparation of tantalum powder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 10 g |
| UN Number | UN3288 |
| Solubility Information | Decomposes in water. Slightly soluble in hydrogen fluoride. |
| Formula Weight | 392.14 |
| Percent Purity | 99.99% |
| Density | 4.560 g/mL |
| Assay | (metals basis) |
| Chemical Name or Material | Potassium heptafluorotantalate (V) |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.