missing translation for 'onlineSavingsMsg'
Learn More

Potassium heptafluorotantalate(V), 99.7%

Product Code. 11333878
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333878 10 g 10g
11323878 50 g 50g
11313878 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333878 Supplier Thermo Scientific Alfa Aesar Supplier No. 040316.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16924-00-8 | F7K2Ta | 392.133 g/mol

It is an important raw material. Used in chemical research. Use as primary and secondary intermediate(Pharmaceutical Intermediates). Used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16924-00-8
Molecular Formula F7K2Ta
Molecular Weight (g/mol) 392.133
MDL Number MFCD00042369
InChI Key DPXAUKPQZOAHGF-UHFFFAOYSA-G
Synonym potassium heptafluorotantalate, potassium tantalum fluoride, dipotassium heptafluorotantalate, tantalum potassium fluoride, potassium fluotantalate, potassium heptafluorotantalate v, potassium fluorotantalate, dipotassium heptafluorotantalum 2-, potassium fluorotanatalate k2taf7, potassium tantalum fluoride k2taf5
PubChem CID 28146
IUPAC Name dipotassium;heptafluorotantalum(2-)
SMILES F[Ta-2](F)(F)(F)(F)(F)F.[K+].[K+]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Decomposes in water; Slightly soluble in HF
Formula Weight 392.14
Percent Purity 99.7%
Density 4.560g/mL
Chemical Name or Material Potassium heptafluorotantalate(V)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.