missing translation for 'onlineSavingsMsg'
Learn More

Potassium formate, 99% (water less than 2%)

Product Code. 11401938
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401938 1000 g 1000g
11411938 250 g 250g
11421938 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11401938 Supplier Thermo Scientific Alfa Aesar Supplier No. A14551.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 590-29-4 | CHKO2 | 84.115 g/mol

Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-29-4
Molecular Formula CHKO2
Molecular Weight (g/mol) 84.115
MDL Number MFCD00013100
InChI Key WFIZEGIEIOHZCP-UHFFFAOYSA-M
Synonym potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
PubChem CID 2735122
ChEBI CHEBI:63316
IUPAC Name potassium;formate
SMILES C(=O)[O-].[K+]

Specifications

Melting Point 165°C to 168°C
Density 1.91
Assay Percent Range 99%
Quantity 1000 g
Beilstein 3623272
Sensitivity Very Hygroscopic
Merck Index 14,7633
Solubility Information Soluble in water and alcohol; Insoluble in ether.
Formula Weight 84.12
Percent Purity 99%
Assay (water <2%)
Chemical Name or Material Potassium Formate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Potassium formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.