missing translation for 'onlineSavingsMsg'
Learn More

Potassium formate, 96%, pract.

Product Code. 15129283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
100g
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15129283 5 kg Plastic Drum 5kg
10677031 100 g Plastic bottle 100g
10563134 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15129283 Supplier Thermo Scientific Acros Supplier No. 418220050
  • ONLINE EXCLUSIVE PRICE
    £281.00
    £263.00 / 5kg
    Save £18.00
    6% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 590-29-4 | CHKO2 | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-29-4
Molecular Formula CHKO2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00013100
InChI Key WFIZEGIEIOHZCP-UHFFFAOYSA-M
Synonym potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
PubChem CID 2735122
ChEBI CHEBI:63316
IUPAC Name potassium;formate
SMILES C(=O)[O-].[K+]

Specifications

Melting Point 165°C to 168°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic Drum
Linear Formula HCOOK
Quantity 5 kg
Merck Index 15,7754
Solubility Information Solubility in water: soluble in water
Formula Weight 84.12
Percent Purity 96%
Grade Practical
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Potassium formate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.