missing translation for 'onlineSavingsMsg'
Learn More

Potassium formate, 96%, pract.

Product Code. 10563134 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
100g
500g
5kg
This item is not returnable. View return policy

Product Code. 10563134

Brand: Thermo Scientific Acros 418225000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-29-4 | CHKO2 | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-29-4
Molecular Formula CHKO2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00013100
InChI Key WFIZEGIEIOHZCP-UHFFFAOYSA-M
Synonym potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
PubChem CID 2735122
ChEBI CHEBI:63316
IUPAC Name potassium;formate
SMILES C(=O)[O-].[K+]

Specifications

Melting Point 165°C to 168°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula HCOOK
Quantity 500 g
Merck Index 15,7754
Solubility Information Solubility in water: soluble in water
Formula Weight 84.12
Percent Purity 96%
Grade Practical
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Potassium formate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.