missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, Puriss., ≥99%, Honeywell

Product Code. 15670540 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 kg
Packaging:
Paper / Poly Bag
Unit Size:
20kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15670540 20 kg Paper / Poly Bag 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15670540 Supplier Honeywell Research Chemicals Supplier No. 0113320KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 7789-23-3
CAS Max % 100.0000%
Quantity 20 kg
Molecular Formula FK
Linear Formula KF
MDL Number MFCD00011398
UN Number UN1812
Beilstein 3902818
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES [F-].[K+]
IUPAC Name potassium fluoride
Molecular Weight (g/mol) 58.10
PubChem CID 522689
ChEBI CHEBI:66872
Formula Weight 58.1g/mol
Percent Purity >99%
Grade puriss.
Packaging Paper / Poly Bag
Chemical Name or Material Potassium fluoride, Puriss., ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.