missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific™

Product Code. 11396977
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11396977

Brand: Thermo Scientific Alfa Aesar 010841.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]

Specifications

Melting Point 41°C
Physical Form Powder
Quantity 10 g
UN Number UN1812
Merck Index 14,7632
Formula Weight 94.13 (58.10 Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Odor Odorless
Refractive Index 1.352
Sensitivity Hygroscopic
Density 2.5g/mL
Assay (metals basis)
Chemical Name or Material Potassium fluoride dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.