missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride Dihydrate, 98%, Reagent grade, Honeywell™

Product Code. p-7120698 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15672160

Brand: Honeywell Research Chemicals 2218725G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]

Specifications

Quantity 5 g
Linear Formula KF · 2H2O
UN Number UN1812
IUPAC Name potassium;fluoride;dihydrate
Formula Weight 94.13g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Fluoride Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.