Product Code. 15672160
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
15672160
5 g
Poly Bottle
5g
15652160
100 g
Poly Bottle
100g
15662160
500 g
Poly Bottle
500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15672160
Supplier
Honeywell Research Chemicals
Supplier No.
2218725G
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Reagent Grade, 98%
≥95% for general laboratory use.
Chemical Identifiers
CAS
13455-21-5
Molecular Formula
FH4KO2
Molecular Weight (g/mol)
94.13
MDL Number
MFCD00149929
InChI Key
MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID
23708904
SMILES
O.O.[F-].[K+]
Specifications
Quantity
5 g
Linear Formula
KF · 2H2 O
UN Number
UN1812
IUPAC Name
potassium;fluoride;dihydrate
Formula Weight
94.13g/mol
Percent Purity
98%
Grade
Reagent
Packaging
Poly Bottle
Chemical Name or Material
Potassium Fluoride Dihydrate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?