missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

Product Code. 11387267
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387267 1 kg 1kg
11377267 25 g 25g
11367267 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11387267 Supplier Thermo Scientific Alfa Aesar Supplier No. 011555.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]

Specifications

Melting Point 41°C
Physical Form Crystalline
Quantity 1 kg
UN Number UN1812
Merck Index 14,7632
Solubility Information Freely soluble in waterSoluble in water; Insoluble in ethanol.
IUPAC Name potassium;fluoride;dihydrate
Formula Weight 94.13 (58.10 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Refractive Index 1.352
Sensitivity Hygroscopic
Density 2.5 g/mL
Chemical Name or Material Potassium fluoride dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.