Learn More
Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals
Description
Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 41°C |
| Physical Form | Crystalline |
| Quantity | 25 g |
| UN Number | UN1812 |
| Merck Index | 14,7632 |
| Solubility Information | Freely soluble in waterSoluble in water; Insoluble in ethanol. |
| IUPAC Name | potassium dihydrate fluoride |
| Formula Weight | 94.13 (58.10 Anhydrous) |
| Percent Purity | ≥98.5% |
| Odor | Odorless |
| Show More |
Safety and Handling
- Potassium fluoride dihydrate
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Potassium fluoride dihydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.