missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, ACS Reagent, ≥99.0%, Honeywell Fluka™

Product Code. p-7120700 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15645410 100 g Poly Bottle 100g
15675410 500 g Poly Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15645410

Brand: Honeywell-Fluka 402931100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 846°C
Color Colorless
Quantity 100 g
Linear Formula KF
UN Number UN1812
Beilstein 3902818
Solubility Information 923g/L at 18°C
Formula Weight 58.1g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly Bottle
Appearance Solid
Density 2.48g/cm3 at 20°C
Chemical Name or Material Potassium fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.