Learn More
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
£24.50 - £83.80
Chemical Identifiers
CAS | 7789-23-3 |
---|---|
Molecular Formula | FK |
Molecular Weight (g/mol) | 58.10 |
MDL Number | MFCD00011398 |
InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
PubChem CID | 522689 |
ChEBI | CHEBI:66872 |
SMILES | [F-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10482201
|
Thermo Scientific Acros
201350250 |
25 g | Plastic bottle |
£24.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10318390
|
Thermo Scientific
201352500 |
250 g | Plastic bottle |
£29.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378430
|
Thermo Scientific
201355000 |
500 g | Plastic bottle |
£30.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10072910
|
Thermo Scientific Acros
201350010 |
1 kg | Plastic bottle |
£47.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10583484
|
Thermo Scientific Acros
201350025 |
2.5 kg | Plastic Drum |
£83.80
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
522689 | |
[F-].[K+] |
FK | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
CHEBI:66872 |
Specifications
1505°C | |
7789-23-3 | |
Crystalline Powder, Crystals and/or Chunks | |
FK | |
MFCD00011398 | |
15, 7753 | |
Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid,insoluble in alcohol unless water is present | |
[F-].[K+] | |
58.10 | |
CHEBI:66872 | |
99% | |
Pass Test | |
Potassium fluoride, 99% |
860°C | |
White | |
99% | |
KF | |
01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium;fluoride | |
522689 | |
58.09 | |
Extra Pure | |
2.48 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-151-5
missing translation for 'rtecsNumber' : TT0700000
missing translation for 'tsca' : TSCA
- Potassium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only