missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11361568
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361568 50 g 50g
11371568 2 g 2g
11381568 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11361568 Supplier Thermo Scientific Alfa Aesar Supplier No. 010980.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.99% (metals basis)
Beilstein 3902818
Solubility Information Freely soluble in water. Insoluble in alcohol.
IUPAC Name potassium;fluoride
Formula Weight 58.1
Refractive Index 1.363
Sensitivity Hygroscopic
Density 2.48 g/mL
Chemical Name or Material Potassium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.