missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferrocyanide trihydrate, 98.5%, ACS reagent

Product Code. 10566412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10566412 5 g Glass bottle 5g
10566602 500 g Plastic bottle 500g
10678144 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10566412 Supplier Thermo Scientific Acros Supplier No. 424130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium hexacyanoferrate(II)
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98.50%
Linear Formula K4Fe(CN)6·3H2O
Merck Index 15, 7752
Solubility Information Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Formula Weight 422.4
Percent Purity 98.5%
Grade ACS Reagent
Packaging Glass bottle
Specific Gravity 1.85
Density 1.8500g/mL
Chemical Name or Material Potassium ferrocyanide trihydrate
Show More Show Less
Product Identifier
  • Potassium ferrocyanide trihydrate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.