missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ethyl Xanthogenate technical grade, 90%, TRC

Product Code. 30424448
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424448 Supplier Toronto Research Chemicals Supplier No. TRCP69710010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Ethyl Xanthogenate technical grade','90%
CAS 140-89-6
Molecular Formula C3 H5 O S2 . K
Quantity 10 g
Synonym Carbonodithioic acid','O-ethyl ester','potassium salt (1:1), Carbonic acid','dithio-','O-ethyl ester','potassium salt (8CI), Carbonodithioic acid','O-ethyl ester','potassium salt (9CI), (O-Ethyl dithiocarbonato)potassium, BKs, BKs (flotation agent), Dithiocarbonic acid O-ethyl ester potassium salt, Ethoxymethanedithioic acid potassium, Ethyl potassium xanthate, Ethyl potassium xanthogenate, Ethylxanthic acid potassium salt, KEX, O-Ethyl dithiocarbonate potassium salt, O-Ethyl potassium dithiocarbonate, Potassium O-ethyl carbonodithioate, Potassium O-ethyl dithiocarbonate, Potassium ethylxanthate, Potassium ethylxanthogenate, Potassium xanthate, Potassium xanthogenate, Z 3, Z 3 (pesticide)
InChI Formula InChI=1S/C3H6OS2.K/c1-2-4-3(5)6;/h2H2,1H3,(H,5,6);/q;+1/p-1
SMILES [K+].CCOC(=S)[S-]
IUPAC Name potassium;ethoxymethanedithioate
Molecular Weight (g/mol) 160.3
Formula Weight 159.9419
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.