missing translation for 'onlineSavingsMsg'
Learn More

Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol, Thermo Scientific Chemicals

Product Code. 11383528
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383528 25 mL 25mL
11393528 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383528 Supplier Thermo Scientific Alfa Aesar Supplier No. 039161.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potassium ethoxide is used widely among others to alkaline varnish degradation . Potassium ethoxide is used as a strong base. It is used to perform transesterification reactions that yield ethyl esters. It is also a suitable base for the malonic ester synthesis. It is also used as Pharmaceutical Intermediates. Other applications include Nano-materials & Thin Films, Engineering & Environmental, Catalysis & Synthesis, Healthcare & Biochemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-58-8
Molecular Formula C2H5KO
Molecular Weight (g/mol) 84.16
MDL Number MFCD00054277
InChI Key RPDAUEIUDPHABB-UHFFFAOYSA-N
Synonym potassium ethoxide, potassium ethanolate, potassium ethylate, ethanol, potassium salt 1:1, potassium ethoxide solution, ethanol, potassium salt, potassiumethoxide, potassium alcohol, etok, koet
PubChem CID 23670592
IUPAC Name potassium;ethanolate
SMILES [K+].CC[O-]

Specifications

Color Colorless to Yellow
Odor Alcohol-like
Linear Formula KOCH2CH3
Quantity 25 mL
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information It reacts vigorously with water.
Formula Weight 84.16
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 15% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Potassium ethoxide
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.