missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium ethoxide, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Potassium ethoxide is used as pharmaceutical and organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 250°C (decomposition) |
Color | Brown to Yellow |
Odor | Alcohol-like |
Linear Formula | KOCH2CH3 |
Quantity | 1 kg |
UN Number | UN3206 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in ethanol, ether |
IUPAC Name | potassium;ethanolate |
Formula Weight | 84.16 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'