missing translation for 'onlineSavingsMsg'
Learn More

Potassium ethoxide, 95%

Product Code. 11357847
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357847 250 g 250g
11377847 50 g 50g
11367847 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357847 Supplier Thermo Scientific Alfa Aesar Supplier No. 014263.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 917-58-8 | C2H5KO | 84.16 g/mol

Potassium ethoxide is used as pharmaceutical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-58-8
Molecular Formula C2H5KO
Molecular Weight (g/mol) 84.16
MDL Number MFCD00054277
InChI Key RPDAUEIUDPHABB-UHFFFAOYSA-N
Synonym potassium ethoxide, potassium ethanolate, potassium ethylate, ethanol, potassium salt 1:1, potassium ethoxide solution, ethanol, potassium salt, potassiumethoxide, potassium alcohol, etok, koet
PubChem CID 23670592
SMILES [K+].CC[O-]

Specifications

Melting Point 250°C (decomposition)
Color Brown to Yellow
Odor Alcohol-like
Linear Formula KOCH2CH3
Quantity 250 g
UN Number UN3206
Sensitivity Air and moisture sensitive
Solubility Information Soluble in ethanol, ether
IUPAC Name potassium ethanolate
Formula Weight 84.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Potassium ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.