missing translation for 'onlineSavingsMsg'
Learn More

Potassium Disulfite, puriss. p.a., Honeywell Fluka™

Product Code. 15613360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
500g
50kg
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15613360 1 kg 1kg
15633360 500 g 500g
15623360 2.5 kg 2.5kg
15653360 6 x 1 kg Pack of 6
15643360 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15613360 Supplier Honeywell-Fluka Supplier No. 312681KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., powder,≥96% (iodometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 1 kg
Linear Formula K2S2O5
UN Number NONH for all modes of transport
Formula Weight 222.32g/mol
Percent Purity ≥96% (iodometric)
Grade puriss. p.a.
Chemical Name or Material Potassium Disulfite
compliance-icons
Product Identifier
  • Potassium disulfite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dipotassium disulphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.