missing translation for 'onlineSavingsMsg'
Learn More

Potassium Disulfate, TRC

Product Code. 30134575
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134575 100 g 100g
30005604 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134575 Supplier Toronto Research Chemicals Supplier No. TRCP695075100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.32
Synonym Potassium Pyrosulfate (K2S2O7), Pyrosulfuric Acid Dipotassium Salt, Dipotassium Disulfate, Dipotassium Pyrosulfate, Potassium Disulfate, Potassium Disulfate (K2S2O7), Potassium Pyrosulfate, Potassium Sulfate (K2S2O7)
IUPAC Name dipotassium;sulfonato sulfate
SMILES [K+].[K+].[O-]S(=O)(=O)OS(=O)(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2K.H2O7S2/c;;1-8(2,3)7-9(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
Formula Weight 253.836
Chemical Name or Material Potassium Disulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.