missing translation for 'onlineSavingsMsg'
Learn More

Potassium diphosphate, 98%, extra pure

Product Code. 10411101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
2.5kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411101 1 kg Plastic bottle 1kg
10378860 25 g Glass Bottle 25g
10454195 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10411101 Supplier Thermo Scientific Acros Supplier No. 269230010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7320-34-5 | K4O7P2 | 330.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7320-34-5
Molecular Formula K4O7P2
Molecular Weight (g/mol) 330.33
MDL Number MFCD00011393
InChI Key RYCLIXPGLDDLTM-UHFFFAOYSA-J
Synonym potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate
PubChem CID 23740
IUPAC Name tetrapotassium;phosphonato phosphate
SMILES [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 1090°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula K4O7P2
Merck Index 15, 7783
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 330.34
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium diphosphate, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.