missing translation for 'onlineSavingsMsg'
Learn More

Potassium dihydrogen phosphate, 98+%

Product Code. p-3345469
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11400113

Brand: Thermo Scientific Alfa Aesar A12142.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Loses H2O at 400°C, forming metaphosphate | CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Loses H{2}O at 400°, forming metaphosphateHygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Quantity 500 g
Assay Percent Range ≥98%
Solubility Information Soluble in water. Slightly soluble in ethanol.
Formula Weight 136.09
Sensitivity Hygroscopic
Density 2.338
Chemical Name or Material Potassium Dihydrogen Phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.