missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dihydrogen Orthophosphate, HPLC for Electrochemical Detection, Fisher Chemical

Product Code. 10598250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
Product Code. Quantity Packaging unitSize
10598250 250 g Plastic powder jar 250g
1 options
This item is not returnable. View return policy

Product Code. 10598250

Brand: Fisher Chemical P/4810/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 253°C
CAS 7778-77-0
Color White
pH 4.5
Physical Form Solid
Quantity 250 g
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.09g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.