missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dihydrogen Orthophosphate, Extra Pure, SLR, Fisher Chemical™

Product Code. 11403753 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Unit Size:
1kg
3kg
500g
5kg
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 11403753

Brand: Fisher Chemical P/4760/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Potassium Dihydrogen Orthophosphate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 5 kg
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.