missing translation for 'onlineSavingsMsg'
Learn More

Potassium dihydrogen citrate hydrate, 99% (dry basis)

Product Code. 11478610
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478610 500 g 500g
11488610 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478610 Supplier Thermo Scientific Alfa Aesar Supplier No. 044415.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 866-83-1 | C6H7KO7 | 230.21 g/mol

Potassium dihydrogen citrate hydrate is used as a buffering agent and a chelating agent. It is used as an alkalizing agent in the renal function. It is found that it increases the bone density, thereby used in research experiments related to the treatment of osteoporosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-83-1
Molecular Formula C6H7KO7
Molecular Weight (g/mol) 230.21
MDL Number MFCD00036472
InChI Key WKZJASQVARUVAW-UHFFFAOYNA-M
Synonym potassium dihydrogen citrate
PubChem CID 73896894
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;potassium
SMILES [K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O

Specifications

Physical Form Chunks and/or Powder
Quantity 500 g
Beilstein 4772973
Merck Index 14,7624
Solubility Information Soluble in water. Slightly soluble in ethanol
Formula Weight 230.22 (Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (dry basis)
Chemical Name or Material Potassium dihydrogen citrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.