missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, TRC

Product Code. 30423383
Change view
Click to view available options
Quantity:
100 mg
1 g
50 g
Unit Size:
100mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423383 50 g 50g
30416718 1 g -
30418396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423383 Supplier Toronto Research Chemicals Supplier No. TRCP69511050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2 O7 . 2 K
Molecular Weight (g/mol) 294.18
Synonym Chromic acid (H2Cr2O7), potassium salt (1:2), Chromic acid (H2Cr2O7), dipotassium salt (9CI), Dichromic acid (H2Cr2O7), dipotassium salt (8CI), C 501, Chromium potassium oxide (K2Cr2O7), Dichromic acid dipotassium salt, Dipotassium bichromate, Dipotassium bichromate (K2Cr2O7), Dipotassium dichromate, Dipotassium dichromate (K2Cr2O7), Dipotassium dichromium heptaoxide, Potassium bichromate, Potassium dichromate, Potassium dichromate (K2(Cr2O7)), Potassium dichromate(VI), SRM 935a
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [K+].[K+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1
Formula Weight 293.7728
Chemical Name or Material Potassium dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.