Learn More
Potassium dichromate, 99+%, ACS reagent, Thermo Scientific Chemicals
£29.00 - £233.00
Chemical Identifiers
CAS | 7778-50-9 |
---|---|
MDL Number | MFCD00011367 |
InChI Key | KMUONIBRACKNSN-UHFFFAOYSA-N |
Synonym | potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
PubChem CID | 24502 |
ChEBI | CHEBI:53444 |
IUPAC Name | dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10371401
|
Thermo Scientific Acros
424110500 |
50 g | Plastic bottle |
£29.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791062
|
Thermo Scientific Acros
424115000 |
500 g | Plastic bottle |
£70.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10475475
|
Thermo Scientific Acros
424110025 |
2.5 kg | Plastic bottle |
£233.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7778-50-9 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Specifications
500.0°C | |
7778-50-9 | |
Crystals | |
Cr2K2O7 | |
MFCD00011367 | |
15, 7748 | |
Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.182 | |
CHEBI:53444 | |
≥99% | |
0.05% max. | |
Potassium dichromate |
398.0°C | |
Orange | |
99+% | |
K2Cr2O7 | |
08,410; 09,383; 10,324; 12,405 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.19 | |
ACS Reagent | |
>500°C |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
Toxic if swallowed.
May intensify fire; oxidizer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Danger
missing translation for 'einecsNumber' : 231-906-6
missing translation for 'rtecsNumber' : HX7680000
missing translation for 'tsca' : TSCA
- Potassium dichromate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 3
- Acute toxicity Category 1
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only