missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, Extra Pure, SLR, Crystals, Fisher Chemical™
Click to view available options
Quantity:
1 kg
5 kg
Specifications
Boiling Point | 500°C |
Melting Point | 398°C |
Color | Orange |
pH | 4 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 294.19g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium dichromate
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 3
- Acute toxicity Category 1
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'