missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, ACROS Organics™

Product Code. 10234412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10234412 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10234412 Supplier Thermo Scientific Acros Supplier No. 382800010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 7778-50-9, 7732-18-5
Quantity 1 L
Molecular Formula Cr2K2O7
MDL Number MFCD00011367
Merck Index 15, 7748
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 294.182
PubChem CID 24502
ChEBI CHEBI:53444
Formula Weight 294.17
Grade Extra Pure
Density 1.01g/mL
Chemical Name or Material Potassium dichromate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.