missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyclohexanebutyrate, AAS, K 18.8%, Thermo Scientific™

Product Code. 11394959
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11394959

Brand: Thermo Scientific Alfa Aesar 087265.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62638-03-3
Molecular Formula C10H17KO2
Molecular Weight (g/mol) 208.34
MDL Number MFCD00013051
InChI Key LBYNWSSEILNGLN-UHFFFAOYSA-M
Synonym potassium cyclohexanebutyrate, potassium 4-cyclohexylbutanoate, potassium 4-cyclohexylbutyrate, cyclohexanebutanoic acid, potassium salt, cyclohexanebutanoic acid, potassium salt 1:1, acmc-20ajuw, cyclohexanebutyric acid potassium salt, cyclohexanebutanoic acid potassium salt, 4-cyclohexylbutanoic acid, potassium salt
PubChem CID 23692296
IUPAC Name potassium;4-cyclohexylbutanoate
SMILES [K+].[O-]C(=O)CCCC1CCCCC1

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 208.33
Grade AAS
Concentration or Composition (by Analyte or Components) K 18.8%
Physical Form Powder
Chemical Name or Material Potassium cyclohexanebutyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.