Product Code. 15661080
Shop All Honeywell Fluka Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
15661080
6 x 1 kg
Pack of 6
15641080
1 kg
1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15661080
Supplier
Honeywell-Fluka
Supplier No.
118136X1KG
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Technical,≥96%
Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90% Variation in color and physical form is possible
Chemical Identifiers
CAS
151-50-8
Molecular Formula
CKN
Molecular Weight (g/mol)
65.116
MDL Number
MFCD00011397
InChI Key
NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID
9032
IUPAC Name
potassium;cyanide
SMILES
[C-]#N.[K+]
Specifications
Quantity
6 x 1 kg
Linear Formula
KCN
UN Number
UN1680
Formula Weight
65.12
Percent Purity
≥96%
Grade
Technical
Chemical Name or Material
Potassium Cyanide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?