missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Cyanide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker ProductsSpecifications
Specifications
| CAS | 151-50-8 |
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Molecular Formula | CKN |
| Synonym | potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french |
| InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
| SMILES | [C-]#N.[K+] |
| IUPAC Name | potassium;cyanide |
| Molecular Weight (g/mol) | 65.116 |
| PubChem CID | 9032 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?