missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog Number 11396439
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]

KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1625°C
Melting Point 634°C
Physical Form Granules
Quantity 250 g
Assay Percent Range 96.0% min.
Beilstein 4652394
Solubility Information Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
Formula Weight 65.12
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.52 g/mL
Chemical Name or Material Potassium cyanide, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'