missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanate, TRC

Product Code. 30409862
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409862 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409862 Supplier Toronto Research Chemicals Supplier No. TRCP69506510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium cyanate
CAS 590-28-3
Molecular Formula C N O . K
Quantity 10 g
Synonym Cyanic acid','potassium salt (1:1) (ACI), Cyanic acid','potassium salt (8CI','9CI), Aero Cyanate, Alicyanate, Bulpur, Isocyanic acid potassium salt, Potassium cyanate, Potassium cyanate (KCNO), Potassium isocyanate
InChI Formula InChI=1S/CHNO.K/c2-1-3;/h3H;/q;+1/p-1
SMILES [K+].[O-]C#N
IUPAC Name potassium;cyanate
Molecular Weight (g/mol) 81.12
Formula Weight 80.9617
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.