missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanate, 97%

Product Code. p-7050024
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11397678

Brand: Thermo Scientific Alfa Aesar 033356.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-28-3 | CKNO | 81.12 g/mol

Potassium cyanate is used in gold mining, organic synthesis and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-28-3
Molecular Formula CKNO
Molecular Weight (g/mol) 81.12
MDL Number MFCD00011411
InChI Key GKKCIDNWFBPDBW-UHFFFAOYSA-M
Synonym potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
PubChem CID 11378442
ChEBI CHEBI:38904
SMILES [K+].[O-]C#N

Specifications

Melting Point 700°C to 900°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Merck Index 14,7625
Solubility Information Soluble in water; Slightly soluble in alcohol
IUPAC Name potassium;cyanate
Formula Weight 81.12
Percent Purity 97%
Odor Odorless
Density 2.056 g/mL
Chemical Name or Material Potassium cyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.