missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanate, 96%, Honeywell Fluka™

Product Code. 15691970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691970 500 g Poly Bottle 500g
15671970 100 g Poly Bottle 100g
15681970 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691970 Supplier Honeywell-Fluka Supplier No. 215074500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.96

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-28-3
Molecular Formula CKNO
Molecular Weight (g/mol) 81.12
MDL Number MFCD00011411
InChI Key GKKCIDNWFBPDBW-UHFFFAOYSA-M
Synonym potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
PubChem CID 11378442
ChEBI CHEBI:38904
SMILES [K+].[O-]C#N

Specifications

Color White
Physical Form Solid
Quantity 500 g
Linear Formula KOCN
UN Number NONH for all modes of transport
Beilstein 3594798
IUPAC Name potassium;cyanate
Formula Weight 81.12g/mol
Percent Purity 96%
Packaging Poly Bottle
Appearance Powder/Chunks
Density 2.056g/mL at 25°C
Chemical Name or Material Potassium Cyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.