missing translation for 'onlineSavingsMsg'
Learn More

Potassium cubes, 99.5% (metals basis), Thermo Scientific™

Catalog Number 15452747
Click to view available options
Quantity:
20 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged in mineral oil
TRUSTED_SUSTAINABILITY
CAS 7440-09-7
Molecular Formula K
MDL Number MFCD00133776
UN Number UN2257
Quantity 20 g
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES [K]
IUPAC Name potassium
Molecular Weight (g/mol) 39.10
PubChem CID 5462222
ChEBI CHEBI:26216
Percent Purity 99.5%
Odor Odorless
Weight ≈20g/cube
Form Cube
Assay (metals basis)
Chemical Name or Material Potassium cubes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'