missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium clavulanate cellulose, MedChemExpress
Description
Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.
Specifications
Specifications
| Chemical Name or Material | Potassium clavulanate cellulose |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C8H9NO5K.(C6H10O5)n |
| Quantity | 100 mg |
| Synonym | Potassium clavulanate:cellulose (1:1) |
| Solubility Information | DMSO : 1 mg/mL (ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](C)[C@@H](CO)O2)[C@@H](CO)O[C@@H](OC)[C@@H]1O.O=C([C@@H](/C(O[C@]3([H])C4)=C/CO)N3C4=O)O[K].[n] |
| Grade | Research |
| Physical Form | Solid |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?