missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, TRC

Product Code. 30423711
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423711 10 g -
30416928 1 g -
30410257 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423711 Supplier Toronto Research Chemicals Supplier No. TRCP69510510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-00-6
Molecular Formula Cr O4 . 2 K
Molecular Weight (g/mol) 194.19
Synonym Chromic acid (H2CrO4), potassium salt (1:2), Chromic acid (H2CrO4), dipotassium salt (8CI,9CI), Bipotassium chromate, Dipotassium chromate, Dipotassium chromate (K2CrO4), Dipotassium chromate(2-), Dipotassium monochromate, Neutral potassium chromate, Potassium chromate, Potassium chromate (K2(CrO4)), Potassium chromate(VI)
IUPAC Name dipotassium;dioxido(dioxo)chromium
SMILES [K+].[K+].[O-][Cr](=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/Cr.2K.4O/q;2*+1;;;2*-1
Formula Weight 193.8476
Chemical Name or Material Potassium chromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.