missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chromate solution, Volumetric, 1/30 M K2CrO4 (0.1N), Solstice
Shop All Honeywell Fluka ProductsDescription
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | K2CrO4 |
| UN Number | UN3082 |
| IUPAC Name | dipotassium;dioxido(dioxo)chromium |
| Formula Weight | 194.19g/mol |
| Grade | Not Available |
| Packaging | Glass bottle |
| Chemical Name or Material | Potassium chromate solution |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?