missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%, Solstice
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Linear Formula | K2CrO4 |
| UN Number | 3288 |
| IUPAC Name | dipotassium;dioxido(dioxo)chromium |
| Formula Weight | 194.19g/mol |
| Percent Purity | ≥99.5% |
| Grade | ACS/puriss. p.a. |
| Density | Vapor Density: 6.7 (vs air) |
| Chemical Name or Material | Potassium Chromate, Reag. Ph. Eur. |
Safety and Handling
Product Identifier
- Potassium chromate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350i-May cause cancer by inhalation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: potassium chromate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?