missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15508084 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15508084 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15508084 Supplier Avantor J.T.Baker Supplier No. 305801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7789-00-6
Physical Form Crystals
Quantity 500 g
Molecular Formula CrK2O4
MDL Number MFCD00011368
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O
IUPAC Name dipotassium;dioxido(dioxo)chromium
Molecular Weight (g/mol) 194.19
PubChem CID 24597
ChEBI CHEBI:75249
Formula Weight 194.19
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.73kg
Chemical Name or Material Potassium Chromate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.