missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate, Certified AR for Analysis, Fisher Chemical™
92.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
92.72 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
92.72 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
CrK2O4, CAS Number-7789-00-6, neutral potassium chromate, dipotassium monochromate, chromate of potass, caswell no. 687, bipotassium chromate, potassium chromate, dipotassium chromate, chromate of potash, potassium chromate vi, tarapacaite, 250g, CHEBI:75249, Yellow, 194.2g/mol
Specifications
Melting Point | 975°C |
Color | Yellow |
pH | 8.6 to 9.8 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 194.2g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium chromate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H340-May cause genetic defects.
- H350i-May cause cancer by inhalation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'