Learn More
Thermo Scientific Chemicals Potassium chromate, ACS, 99.0% min
Description
In finishing of leather, rust proofing of metals, textile mordant, as an analytical lab reagent. Potassium chromate enjoys a broad assortment of industrial purposes. In the fabric industry, manufacturers use potassium chromate to tan leather and dye clothing items and materials. In science, potassium chromate is often used as an indicator, meaning it can identify the presence, location and quantity of certain elements such as iron. Beyond these applications, potassium chromate also functions as an oxidizing agent in manufacturing and scientific experiments. Some nuclear power plants currently use substances other than potassium chromate to inhibit corrosion in closed cooling-water systems. Potassium chromate is applied as a chemical indicator of chlorine ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 975°C |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.0% min. |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 194.19 |
Grade | ACS |
Odor | Odorless |
Appearance | Yellow crystals |
Show More |
RUO – Research Use Only