Learn More
Potassium chloride, ultra dry, 99.95% (metals basis)
Description
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia (a condition in which the level of potassium will be low in blood) with its pharmaceutical preparation. Furthermore, it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1500°C (sublimation) |
| Melting Point | 773°C |
| Color | White |
| Physical Form | Beads |
| Quantity | 50 g |
| Assay Percent Range | 99.95% (metals basis) |
| Solubility Information | Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether. |
| Formula Weight | 74.55 |
| Odor | Odorless |
| Refractive Index | 1.49 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.