missing translation for 'onlineSavingsMsg'
Learn More

3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™
BIOPHARMA

Product Code. 11503172 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11503172 500 mL 500mL
11593162 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11503172 Supplier Reagecon Supplier No. EFS3AC5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for refilling or replacing the reference electrolyte in silver/silver chloride or calomel reference electrodes. 3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™ is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements.

A good electrolyte must fulfil a number of conditions: the equitransference of the cation/anion combination should be as close as possible to being equimobile, have constant chloride activity, be of high electrical conductance and as non-chemically reactive as possible.

Potassium Chloride (KCl) fulfils all of these conditions to a greater or lesser extent, and is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements where silver/silver chloride or calomel reference electrodes are used. 3M KCl is appropriate where the working temperature is at least 20°C.

Chemical Identifiers

CAS 7447-40-7 , 7783-90-6
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Concentration 3 mol/L, Saturated with AgCl
Physical Form Liquid
Quantity 500 mL
Solution Type Electrode Filling
IUPAC Name potassium;chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.