missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Solution 4M, Pure, Electrode Filling Solution with Silver Chloride, Fisher Chemical

Product Code. 10070180 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Packaging:
Amber glass bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10070180

Brand: Fisher Chemical J/2984/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7447-40-7
Color Colorless
Physical Form Liquid
Quantity 100 mL
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Packaging Amber glass bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.